missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRBP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10386-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
May be involved in transcriptional regulation.Especificaciones
KRBP | |
Polyclonal | |
Unconjugated | |
ZNF426 | |
K RBP, KRBP, zinc finger protein 426, ZNF426 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
235028, 79088 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8R1D1, Q9BUY5 | |
ZNF426 | |
KRBP Fusion Protein Ag0585 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |