missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KLK3 (Human) Recombinant Protein
Human KLK3 full-length ORF ( AAH05307, 1 a.a. - 261 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00000354-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MWVPVVFLTLSVTWIGAAPLILSRIVGGWECEKHSQPWQVLVASRGRAVCGGVLVHPQWVLTAAHCIRNKSVILLGRHSLFHPEDTGQVFQVSHSFPHPLYDVSLLKNRFLRPGDDSSHDLMLLRLSEPAELTDAVKVMDLPTQEPALGTTCYASGWGSIEPEEFLTPKKLQCVDLHVISNDVCAQVHPQKVTKLMLCAGRWTGGKSTCSGDSGGPLVCNGVLQGITSWGSEPCALPERPSLYTKVVHYRKWIKDTIVANP
Especificaciones
AAH05307 | |
kallikrein-related peptidase 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KLK3 | |
GST |
354 | |
Wheat germ expression system | |
25 μg | |
APS, KLK2A1, PSA, hK3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |