missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KCNJ12 (Human) Recombinant Protein
Human KCNJ12 full-length ORF ( AAH27982, 1 a.a. - 433 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00003768-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MTAASRANPYSIVSSEEDGLHLVTMSGANGFGNGKVHTRRRCRNRFVKKNGQCNIEFANMDEKSQRYLADMFTTCVDIRWRYMLLIFSLAFLASWLLFGIIFWVIAVAHGDLEPAEGRGRTPCVMQVHGFMAAFLFSIETQTTIGYGLRCVTEECPVAVFMVVAQSIVGCIIDSFMIGAIMAKMARPKKRAQTLLFSHNAVVALRDGKLCLMWRVGNLRKSHIVEAHVRAQLIKPRVTEEGEYIPLDQIDIDVGFDKGLDRIFLVSPITILHEIDEASPLFGISRQDLETDDFEIVVILEGMVEATAMTTQARSSYLANEILWGHRFEPVLFEEKNQYKIDYSHFHKTYEVPSTPRCSAKDLVENKFLLPSANSFCYENELAFLSRDEEDEADGDQDGRSRDGLSPQARHDFDRLQAGGGVLEQRPYRRESEI
Especificaciones
AAH27982 | |
potassium inwardly-rectifying channel, subfamily J, member 12 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KCNJ12 | |
GST |
3768 | |
Wheat germ expression system | |
25 μg | |
FLJ14167, IRK2, KCNJN1, Kir2.2, Kir2.2v, hIRK, hIRK1, hkir2.2x, kcnj12x | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |