missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KCNG1 (Human) Recombinant Protein
Human KCNG1 full-length ORF ( AAH06367, 1 a.a. - 275 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00003755-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MTLLPGDNSDYDYSALSCTSDASFHPAFLPQRQAIKGAFYRRAQRLRPQDEPRQGCQPEDRRRRIIINVGGIKYSLPWTTLDEFPLTRLGQLKACTNFDDILNVCDDYDVTCNEFFFDRNPGAFGTILTFLRAGKLRLLREMCALSFQEELLYWGIAEDHLDGCCKRRYLQKIEEFAEMVEREEEDDALDSEGRDSEGPAEGEGRLGRCMRRLRDMVERPHSGLPGKVFACLSVLFVTVTAVNLSVSTLPSLREEEEQVRAHAPRGNAPPRGKGL
Especificaciones
AAH06367 | |
potassium voltage-gated channel, subfamily G, member 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KCNG1 | |
GST |
3755 | |
Wheat germ expression system | |
25 μg | |
K13, KCNG, KV6.1, MGC12878, kH2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |