missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human JAK2, GST Tag Recombinant Protein
JAK2 Recombinant Human Protein
Marca: Invitrogen™ PV4288
Detalles adicionales : Peso : 0.00945kg
Descripción
Janus kinase 2 (JAK2) is a protein tyrosine kinase involved in a specific subset of cytokine receptor signaling pathways and is mutated in polycythemia vera. This kinase contains the catalytic domain (JH1) of JAK2.Especificaciones
NP_004963 | |
Protein Assays and Analysis, Kinase Assay | |
66.6kDa | |
1mg | |
For research use only. Not for use in diagnostic procedures | |
JAK2 | |
JAK2 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL Abl1 peptide substrate (EAIYAAPFAKKK) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 808-1132 | |
Recombinant human protein, catalytic domain (amino acids 808-1132), GST-tagged, expressed in insect cells containing additional C-terminal amino acids (legacy from Gateway™ cloning). Activated in vitro via autophosphorylation | |
GST Tag | |
Frozen |
0.29mg/mL | |
Frozen | |
JAK2 Recombinant Human Protein | |
–80°C | |
1 Each | |
JAK2 Recombinant Human Protein | |
JAK | |
JAK2 Recombinant Human Protein | |
Human | |
Baculovirus | |
95% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.