missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISYNA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14142-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Myoinositol, the most common naturally occurring form of inositol, is a component of plasma membrane phospholipids and functions as a cell signaling molecule. ISYNA1 (EC 5.5.1.4), or IPS, is a rate-limiting enzyme that catalyzes the de novo synthesis of myoinositol 1-phosphate from glucose 6-phosphate. Myoinositol, the most common naturally occurring form of inositol, is a component of plasma membrane phospholipids and functions as a cell signaling molecule. ISYNA1 (EC 5.5.1.4), or IPS, is a rate-limiting enzyme that catalyzes the de novo synthesis of myoinositol 1-phosphate from glucose 6-phosphate (Seelan et al., 2004 ).Especificaciones
ISYNA1 | |
Polyclonal | |
Unconjugated | |
ISYNA1 | |
hINO1, hIPS, Ino1, INOS, IPS, IPS 1, ISYNA1, MI 1 P synthase, MIP synthase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
290651, 51477, 71780 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6AYK3, Q9JHU9, Q9NPH2 | |
Isyna1 | |
ISYNA1 Fusion Protein Ag5272 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |