missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF6 Monoclonal Antibody (06), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA543524
Detalles adicionales : Peso : 0.01000kg
Descripción
Specificity: Human IRF6
This gene encodes a member of the interferon regulatory transcription factor (IRF) family. Family members share a highly-conserved N-terminal helix-turn-helix DNA-binding domain and a less conserved C-terminal protein-binding domain. Mutations in this gene can cause van der Woude syndrome and popliteal pterygium syndrome. This protein is involved in palate formation.Especificaciones
IRF6 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O14896 | |
Irf6 | |
A synthetic peptide corresponding to the C-terminus of the human IRF6 | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG3 |
Immunocytochemistry | |
06 | |
Unconjugated | |
Irf6 | |
AI876454; E230028I05Rik; interferon regulatory factor 6; IRF6; IRF-6; irf6 protein; IRF-6; transcription factor; LPS; mirf6; OFC6; PIT; PPS; PPS1; transcriptional factor; VWS; VWS1; zgc:63500 | |
Mouse | |
Protein A | |
RUO | |
3664 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |