missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF4 Mouse anti-Human, Porcine, Rat, Clone: 1A12D5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66451-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The IRF (interferon regulatory factor) family of transcription factors are characterized by an unique tryptophan pentad repeat DNA-binding domain. The IRFs are important in the regulation of interferons in response to infection by virus, and in the regulation of interferon-inducible genes. IRF4 is lymphocyte specific and negatively regulates toll-like-receptor (TLR) signaling that is central to the activation of innate and adaptive immune systems. A chromosomal translocation involving this gene and the IgH locus, t(6;14)(p25;q32), may be a cause of multiple myeloma. Alternatively spliced transcript variants have been found.Especificaciones
IRF4 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15306 | |
Irf4 | |
IRF4 Fusion Protein Ag25203 | |
20 μL | |
Primary | |
Porcine, Rat, Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1A12D5 | |
Unconjugated | |
IRF4 | |
interferon regulatory factor 4, IRF 4, IRF4, LSIRF, Multiple myeloma oncogene 1, MUM1, NF EM5 | |
Mouse | |
Protein G | |
RUO | |
100144625, 291100, 3662 | |
-20°C | |
Liquid |