missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IRAK1, GST Tag Recombinant Protein
IRAK1 Recombinant Human Protein
Marca: Invitrogen™ PV4403
Detalles adicionales : Peso : 0.00900kg
Descripción
IL-1 receptor associated kinases (IRAKs) are serine⁄threonine kinases involved in signaling via Toll⁄IL-1 pathways.Especificaciones
NP_001560 | |
83.4 kDa | |
IRAK1, IRAK, pelle | |
aa 194-712 | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: TKL Group | |
IRAK1 | |
Baculovirus | |
Recombinant, Catalytic | |
Serine/Threonine Kinases |
Kinase Assay | |
10μg | |
IRAK Kinase Family | |
Human | |
GST Tag | |
Biochemical binding/kinetic | |
Dry Ice | |
interleukin-1 receptor-associated kinase 1 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.