Learn More
Invitrogen™ TOPO™ TA Cloning™ Kit, with One Shot™ TOP10F' Chemically Competent E. coli, no manual
Designed for cloning PCR products directly from a PCR reaction in just 5 minutes
Marca: Invitrogen™ KNM455001
Detalles adicionales : Peso : 0.00500kg
Descripción
TOPO TA Cloning kits• Alta eficiencia
• 'One-step'
Especificaciones
Kit, Cloning | |
TOP10F´ | |
25 Reactions |
Box 1: Topoisomerase I-activated pCR2.1-TOPO vector, PCR buffer, Salt solution, dNTPs, Control template, M13 forward and reverse primers, Control PCR primers, Sterile water. Box 2: One Shot Chemically Competent or Electrocomp E. coli, S.O.C. medium, Supercoiled pUC19 control plasmid | |
Cloning, PCR Cloning |
For Research Use Only. Not for use in diagnostic procedures.