Learn More
Integrin beta-6 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 28378-1-AP-20UL
Detalles adicionales : Peso : 0.00020kg
 Ver más versiones de este producto
Código de producto. 17211406
  • 129.00€ / 20 microlitros
Fecha estimada de envío: 23-12-2022
para ver el stock
Descripción
Descripción
The Integrin beta-6 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human Integrin beta-6. This antibody recognizes human, mouse antigen. The Integrin beta-6 antibody has been validated for the following applications: IHC, WB,ELISA analysis.
Especificaciones
Especificaciones
Integrin beta-6
Polyclonal
Unconjugated
20 μL
3694
Liquid
ELISA, Immunohistochemistry, Western Blot
1000 μg/mL
Rabbit
Primary
Mouse, Human