missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10905-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody is specifically against p47ING3.
ING3 belongs to a family of proteins containing the plant homeodomain (PHD) finger, which includes transcription factors and proteins that regulate chromatin structure. ING3 is a component of the NuA4 histone acetyltransferase (HAT) complex and can activate p53 trans-activated promoters, including promoters of p21/waf1 and bax. Overexpression of ING3 has been shown to inhibit cell growth and induce apoptosis. Allelic loss and reduced expression of the ING3 gene were detected in head and neck cancers.Especificaciones
ING3 | |
Polyclonal | |
Unconjugated | |
ING3 | |
Eaf4, ING2, ING3, Inhibitor of growth protein 3, p47ING3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54556, 71777 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8VEK6, Q9NXR8 | |
ING3 | |
ING3 Fusion Protein Ag1357 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |