Learn More
IgG2b Rat, FITC, Clone: RTK4530, Isotype Control, STEMCELL Technologies
Rat Monoclonal Antibody
Marca: Stemcell Technologies 60077FI.1
Detalles adicionales : Peso : 0.00110kg
Descripción
- The RTK4530 antibody (IgG2b, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody
- Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used
- If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen
- The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components
- This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining)
Especificaciones
IgG2b | |
Monoclonal | |
FITC | |
50 μg | |
IgG2b κ |
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
RTK4530 | |
Rat | |
Isotype Control (Primary) | |
Other |