Learn More
IgG2b Rat, FITC, Clone: RTK4530, Isotype Control, STEMCELL Technologies
Rat Monoclonal Antibody
Marca:  Stemcell Technologies 60077FI.2
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17108921
  • 201.15€ / 100 tests
Fecha estimada de envío: 24-05-2024
para ver el stock
Añadir a la cesta
Descripción
Descripción
  • The RTK4530 antibody (IgG2b, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody
  • Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used
  • If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen
  • The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components
  • This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining)
Especificaciones
Especificaciones
IgG2b
Monoclonal
FITC
100 Tests
IgG2b κ
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
RTK4530
Rat
Isotype Control (Primary)
Other