Learn More
IgG light chain (lambda) Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11541-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Immunoglobulin classes share the same basic four polypeptide chain structure of two heavy chains (five types) and two light chains (kappa, lambda; both having a molecular weight of 22.5 kDa). Kappa and lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region. The ratio of kappa to lambda is 70:30. Antibodies to the kappa light chain are reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin′s lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore maligt.Especificaciones
IgG light chain (lambda) | |
Polyclonal | |
Unconjugated | |
IGL | |
IGL | |
IgG light chain (lambda) Fusion Protein Ag2064 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
IGL, IGL-, immunoglobulin lambda locus | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3535 | |
-20°C | |
Liquid |