Learn More
Epredia™ IgE (ε -Heavy Chain), Rabbit, Polyclonal, Epredia™
IgE (ε-Heavy Chain), Rb PAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Marca: Epredia™ RB-1435-A0
Detalles adicionales : Peso : 0.00170kg
Descripción
Research Areas: Immunoglobulins; The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.Host Species: Rabbit
Clone: Polyclonal
Isotype: IgG
Species Reactivity: Human. Others not known.
Immunogen: IgE isolated from a pool of normal human sera
Molecular Weight: 50kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Especificaciones
IgE (epsilon-Heavy Chain) | |
Polyclonal | |
Unconjugated | |
IgE isolated from a pool of normal human sera | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
Polyclonal | |
Rabbit | |
100 μL | |
Immunoglobulins | |
Human | |
IgG |