missing translation for 'onlineSavingsMsg'
                
Learn More
Learn More
IgD, Rat anti-Mouse, APC-H7, Clone: 11-26c.2a, BD Pharmingen™
Rat Monoclonal Antibody
Marca: BD Biosciences 565348
                        
                            
                        
                        
                        
                            
                        
                        
                        
                        
                            
                                    
                                Este artículo no se puede devolver.
                                Vea la política de devoluciones
                                
                        
                    
                Descripción
IgD Monoclonal antibody specifically detects IgD in Mouse samples. It is validated for Flow cytometry.
            Especificaciones
| IgD | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Cell Biology | |
| Mouse | |
| Purified | 
| Flow Cytometry | |
| 11-26c.2a | |
| APC-H7 | |
| IGHD; Igh-5; Immunoglobulin heavy chain 5; Ig delta chain C region | |
| Not reported | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with APC-H7 under optimum conditions, and unconjugated antibody and APC-H7 were removed. | |
| IgG2a κ | 
                    Corrección del contenido de un producto
                
                Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
			IgD, Rat anti-Mouse, APC-H7, Clone: 11-26c.2a, BD Pharmingen™ > 50μg; APC-H7
            For Reseach Use Only.
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido