missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFNGR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10808-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene (IFNGR1) encodes the ligand-binding chain (alpha) of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. A genetic variation in IFNGR1 is associated with susceptibility to Helicobacter pylori infection. In addition, defects in IFNGR1 are a cause of mendelian susceptibility to mycobacterial disease, also known as familial disseminated atypical mycobacterial infection.Especificaciones
IFNGR1 | |
Polyclonal | |
Unconjugated | |
IFNGR1 | |
CD119, CDw119, IFN gamma R1, IFN gamma receptor 1, IFNGR, IFNGR1, interferon gamma receptor 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3459 | |
-20°C | |
Liquid |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P15260 | |
IFNGR1 | |
IFNGR1 Fusion Protein Ag1114 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |