missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFITM1 Recombinant Rabbit Monoclonal Antibody (001), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen MA543443
Detalles adicionales : Peso : 0.01000kg
Descripción
Specificity: Human IFITM1
IFITM1 (Interferon inducible transmembrane protein 1) is a member of the IFN-inducible transmembrane protein family. It is an essential mediator of interferon-gamma-induced antiproliferation and plays a role in the control of cell growth. IFITM1 is upregulated in several tumor types and may be useful as a tumor biomarker. Both mouse IFITM1 and IFITM3 are expressed on the cell surfaces of primordial germ cells in a developmentally-regulated manner. IFITM1 activity is required for primordial germ cell transit, and IFITM1 acts as a repulsive molecule by repelling non-IFITM1-expressing primordial germ cells from the mesoderm into the endoderm.Especificaciones
IFITM1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P13164 | |
Ifitm1 | |
A synthetic peptide corresponding to the center region of the Human IFITM1 | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
001 | |
Unconjugated | |
Ifitm1 | |
1110036C17Rik; 27-Sep; 9-27; CD225; dispanin subfamily A member 2a; DSPA2a; Fragilis protein 2; fragilis2; IFI17; IFITM1; ifitm-like protein 2; interferon induced transmembrane protein 1; interferon induced transmembrane protein 2 like; interferon-induced protein 17; Interferon-induced transmembrane protein 1; interferon-inducible protein 9-27; LEU13; Leu-13 antigen; Mil2; Mil-2; Mouse ifitm-like protein 2 | |
Rabbit | |
Protein A | |
RUO | |
8519 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |