missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDI2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16701-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Catalyzes the 1,3-allylic rearrangement of the homoallylic substrate isopentenyl (IPP) to its highly electrophilic allylic isomer, dimethylallyl diphosphate (DMAPP).Especificaciones
IDI2 | |
Polyclonal | |
Unconjugated | |
IDI2 | |
IDI2, IPP isomerase 2, IPPI2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
91734 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BXS1 | |
IDI2 | |
IDI2 Fusion Protein Ag10098 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |