missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICMT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 51001-2-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes the third of three enzymes that posttranslationally modify isoprenylated C-terminal cysteine residues in certain proteins and target those proteins to the cell membrane. This enzyme localizes to the endoplasmic reticulum. Alternative splicing may result in other transcript variants, but the biological validity of those transcripts has not been determined.Especificaciones
ICMT | |
Polyclonal | |
Unconjugated | |
ICMT | |
HSTE14, ICMT, MST098, MSTP098, PCCMT, PCMT, PPMT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170818, 23463, 57295 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60725, Q9EQK7, Q9WVM4 | |
Icmt | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |