Learn More
STEMCELL Technologies Hydrocortisone Stock Solution
Hydrocortisone Stock Solution (200X) for use with PneumaCult™-ALI, MammoCult™, EpiCult™, 10 pack
Marca:  STEMCELL Technologies 07925
Detalles adicionales : Peso : 0.01330kg
 Ver más versiones de este producto
Código de producto. 17199911
  • 52.50€ / 3 ml
Fecha estimada de envío: 25-04-2024
para ver el stock
Descripción
Descripción
  • Hydrocortisone Stock Solution is recommended for use as a cell culture supplement and for the preparation of PneumaCult™, MammoCult™, and EpiCult™ media when culturing airway epithelial cells, mammary primary cells, or tumor cell lines
  • Hydrocortisone Stock Solution is provided at a concentration of 96 μg/mL, which is ideal as a 200X stock solution for use with PneumaCult™-ALI (Catalog #05001), MammoCult™ (Catalog #05620), EpiCult™-B (Human; Catalog #05601), and EpiCult™-C (Catalog #05630) media
  • It may also be used for preparing PneumaCult™-Ex Medium (Catalog #05008) or PneumaCult™-Ex Plus Medium (Catalog #05040)
Especificaciones
Especificaciones
Airway Cells, Cancer Cells and Cell Lines, Mammary Cells
Cell Culture
3 mL