missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZSCAN21 Partial ORF (NP_666019, 136 a.a. - 224 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007589-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: TPPNEQKPVWEKISSSGTAKESPSSMQPQPLETSHKYESWGPLYIQESGEEQEFAQDPRKVRDCRLSTQHEESADEQKGSEAEGLKGDIEspecificaciones
NP_666019 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.42kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
TPPNEQKPVWEKISSSGTAKESPSSMQPQPLETSHKYESWGPLYIQESGEEQEFAQDPRKVRDCRLSTQHEESADEQKGSEAEGLKGDI | |
RUO | |
ZSCAN21 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7589 | |
ZSCAN21 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434L134/DKFZp686H10254/NY-REN-21/ZNF38/Zipro1 | |
ZSCAN21 | |
Recombinant | |
wheat germ expression system |