missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF85 Partial ORF (NP_003420, 2 a.a. - 76 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007639-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: GPLTFRDVAIEFSLKEWQCLDTAQRNLYRNVMLENYRNLVFLGITVSKPDLITCLEQGKEAWSMKRHEIMVAKPTEspecificaciones
NP_003420 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.99kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GPLTFRDVAIEFSLKEWQCLDTAQRNLYRNVMLENYRNLVFLGITVSKPDLITCLEQGKEAWSMKRHEIMVAKPT | |
RUO | |
ZNF85 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7639 | |
ZNF85 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HPF4/HTF1/MGC78566 | |
ZNF85 | |
Recombinant | |
wheat germ expression system |