missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF224 Partial ORF (NP_037530.1, 95 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007767-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: HQEWSFQQIWEKIASDLTRSQDLVINSSQFSKEGDFPCQTEAGLSVIHTRQKSSQGNGYKPSFSDVSHFDFHQQLHSGEKSHTCDECGKNFCYISAEspecificaciones
NP_037530.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.3kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HQEWSFQQIWEKIASDLTRSQDLVINSSQFSKEGDFPCQTEAGLSVIHTRQKSSQGNGYKPSFSDVSHFDFHQQLHSGEKSHTCDECGKNFCYISA | |
RUO | |
ZNF224 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7767 | |
ZNF224 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BMZF-2/BMZF2/KOX22/ZNF255/ZNF27 | |
ZNF224 | |
Recombinant | |
wheat germ expression system |