Learn More
Abnova™ Human ZIC4 Partial ORF (NP_115529, 249 a.a. - 319 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00084107-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. Members of this family are important during development, and have been associated with X-linked visceral heterotaxy and holoprosencephaly type 5. This gene is closely linked to the gene encoding zinc finger protein of the cerebellum 1, a related family member on chromosome 3. This gene encodes a protein of unknown function. [provided by RefSeq]
Sequence: SDRKKHSHVHTSDKPYTCKVRGCDKCYTHPSSLRKHMKVHGRSPPPSSGYDSATPSALVSPSSDCGHKSQVEspecificaciones
NP_115529 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.55kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SDRKKHSHVHTSDKPYTCKVRGCDKCYTHPSSLRKHMKVHGRSPPPSSGYDSATPSALVSPSSDCGHKSQV | |
RUO | |
ZIC4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
84107 | |
ZIC4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ42609/FLJ45833 | |
ZIC4 | |
Recombinant | |
wheat germ expression system |