missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZFP90 Full-length ORF (AAH33165.1, 1 a.a. - 224 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00146198-P01.10ug
Informations supplémentaires : Peso : 0.00010kg
Description
Sequence: MRIHKRGKPYQSSNYSIDFKHSTSLTQDESTLTEVKSYHCNDCGEDFSHITDFTDHQRIHTAENPYDCEQAFSQQAISHPGEKPYQCNVCGKAFKRSTSFIEHHRIHTGEKPYECNECGEAFSRRSSLTQHERTHTGEKPYECIDCGKAFSQSSSLIQHERTHTGEKPYECNECGRAFRKKTNLHDHQRIHTGEKPYSCKECGKNFSRSSALTKHQRIHTRNKLSpécification
AAH33165.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
52.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NK10/ZNF756 | |
ZFP90 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
146198 | |
ZFP90 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRIHKRGKPYQSSNYSIDFKHSTSLTQDESTLTEVKSYHCNDCGEDFSHITDFTDHQRIHTAENPYDCEQAFSQQAISHPGEKPYQCNVCGKAFKRSTSFIEHHRIHTGEKPYECNECGEAFSRRSSLTQHERTHTGEKPYECIDCGKAFSQSSSLIQHERTHTGEKPYECNECGRAFRKKTNLHDHQRIHTGEKPYSCKECGKNFSRSSALTKHQRIHTRNKL | |
RUO | |
ZFP90 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |