Learn More
Abnova™ Human ZBTB46 Full-length ORF (AAH52269.1, 1 a.a. - 589 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca:  Abnova™ H00140685-P01.10ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 16168223
  • 335.00€ / 10 microgramos
Fecha estimada de envío: 12-06-2024
para ver el stock
Descripción
Descripción
Sequence: MNNRKEDMEIASHYRHLLRELNEQRQHGVLCDVCVVVEGKVFKAHKNVLLGSSRYFKTLYCQVQKTSEQATVTHLDIVTAQGFKAIIDFMYSAHLALTSRNVIEVMSAASFLQMTDIVQACHDFIKAALDISIKSDASDELAEFEIGASSSSSTEALISAVMAGRSISPWLARRTSPANSSGDSAIASCHDGGSSYGKEDQEPKADGPDDVSSQPLWPGDVGYGPLRIKEEQVSPSQYGGSELPSAKDGAVQNSFSEQSAGDAWQPTGRRKNRKNKETVRHITQQVEDDSRASSPVPSFLPTSGWPFSSRDSNADLSVTEASSSDSRGERAELYAQVEEGLLGGEASYLGPPLTPEKDDALHQATAVANLRAALMSKNSLLSLKADVLGDDGSLLFEYLPRGAHSLSLNEFTVIRKKFKCPYCSFSAMHQCILKRHMRSHTGERPYPCEICGKKFTRREHMKRHTLVHSKDKKYVCKVCSRVFMSAASVGIRHGSRRHGVCTDCAGRGMAGPLDHGGGGGEGSPEALFPGDGPYLEDPEDPRGEAEELGEDDEGLAPEDALLADDKDEEDSPRPRSPPGGPDKDFAWLS
Especificaciones
Especificaciones
AAH52269.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
90.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BTBD4/FLJ13502/RINZF/ZNF340/dJ583P15.7/dJ583P15.8
ZBTB46
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
140685
ZBTB46 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNNRKEDMEIASHYRHLLRELNEQRQHGVLCDVCVVVEGKVFKAHKNVLLGSSRYFKTLYCQVQKTSEQATVTHLDIVTAQGFKAIIDFMYSAHLALTSRNVIEVMSAASFLQMTDIVQACHDFIKAALDISIKSDASDELAEFEIGASSSSSTEALISAVMAGRSISPWLARRTSPANSSGDSAIASCHDGGSSYGKEDQEPKADGPDDVSSQPLWPGDVGYGPLRIKEEQVSPSQYGGSELPSAKDGAVQNSFSEQSAGDAWQPTGRRKNRKNKETVRHITQQVEDDSRASSPVPSFLPTSGWPFSSRDSNADLSVTEASSSDSRGERAELYAQVEEGLLGGEASYLGPPLTPEKDDALHQATAVANLRAALMSKNSLLSLKADVLGDDGSLLFEYLPRGAHSLSLNEFTVIRKKFKCPYCSFSAMHQCILKRHMRSHTGERPYPCEICGKKFTRREHMKRHTLVHSKDKKYVCKVCSRVFMSAASVGIRHGSRRHGVCTDCAGRGMAGPLDHGGGGGEGSPEALFPGDGPYLEDPEDPRGEAEELGEDDEGLAPEDALLADDKDEEDSPRPRSPPGGPDKDFAWLS
RUO
ZBTB46