Learn More
Abnova™ Human XRN1 Full-length ORF (AAH48104.1, 1 a.a. - 459 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00054464-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
SEP1 localizes to cytoplasmic foci containing a complex of mRNA-degrading enzymes. In addition to mRNA metabolism, yeast Sep1 has been implicated in a variety of nuclear and cytoplasmic functions, including homologous recombination, meiosis, telomere maintenance, and microtubule assembly.[supplied by OMIM]
Sequence: MGVPKFYRWISERYPCLSEVVKEHQIPEFDNLYLDMNGIIHQCSHPNDDDVHFRISDDKIFTDIFHYLEVLFRIIKPRKVFFMAVDGVAPRAKMNQQRGRRFRSAKEAEDKIKKAIEKGETLPTEARFDSNCITPGTEFMAKLHEHLKYFVNMKISTDKSWQGVTIYFSGHETPGEGEHKIMEFIRSEKAKPDHDPNTRHCLYGLDADLIMLGLTSHEAHFSLLREEVRFGGKKTQRVCAPEETTFHLLHLSLMREYIDYEFSVLKEKITFKYDIERIIDDWILMGFLVGNDFIPHLPHLHINHDALPLLYGTYVTILPELGGYINESGHLNLPRFEKYLVKLSDFDREHFSEVFVDLKWFESKVGNKYLNEAAGVAAEEARNYKEKKKLKGQENSLCWTALDKNEGEMITSKDNLEDETEDDDLFETEFRQYKRTYYMTKMGVDVVSEYVFANAFILKEspecificaciones
AAH48104.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
80.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434P0721/DKFZp686B22225/DKFZp686F19113/FLJ41903/SEP1 | |
XRN1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54464 | |
XRN1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGVPKFYRWISERYPCLSEVVKEHQIPEFDNLYLDMNGIIHQCSHPNDDDVHFRISDDKIFTDIFHYLEVLFRIIKPRKVFFMAVDGVAPRAKMNQQRGRRFRSAKEAEDKIKKAIEKGETLPTEARFDSNCITPGTEFMAKLHEHLKYFVNMKISTDKSWQGVTIYFSGHETPGEGEHKIMEFIRSEKAKPDHDPNTRHCLYGLDADLIMLGLTSHEAHFSLLREEVRFGGKKTQRVCAPEETTFHLLHLSLMREYIDYEFSVLKEKITFKYDIERIIDDWILMGFLVGNDFIPHLPHLHINHDALPLLYGTYVTILPELGGYINESGHLNLPRFEKYLVKLSDFDREHFSEVFVDLKWFESKVGNKYLNEAAGVAAEEARNYKEKKKLKGQENSLCWTALDKNEGEMITSKDNLEDETEDDDLFETEFRQYKRTYYMTKMGVDVVSEYVFANAFILK | |
RUO | |
XRN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |