missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human WDFY3 Full-length ORF (AAH15214, 1 a.a. - 420 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00023001-P01.10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a protein which contains WD repeats and an FYVE domain. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been defined. [provided by RefSeq]
Sequence: MGTSKEKAKTATLKQALLGHTDTVTCATASLAYHIIVSGSRDRTCIIWDLNKLSFLTQLRGHRAPVSALCINELTGDIVSCAGTYIHVWSINGNPIVSVNTFTGRSQQIICCCMSEMNEWDTQNVIVTGHSDGVVRFWRMEFLQVPETPAPEPAEVLEMQEDCPEAQIGQEAQDEDSSDSEADEQSISQDPKDTPSQPSSTSHRPRAASCRATAAWCTDSGSDDSRRWSDQLSLDEKDGFIFVNYSEGQTRAHLQGPLSHPHPNPIEVRNYSRLKPGYRWERQLVFRSKLTMHTAFDRKDNAHPAEVTALGISKDHSRILVGDSRGRVFSWSVSDQPGRSAADHWVKDEGGDSCSGCSVRFSLTERRHHCRNCGQLFCQKCSRFQSEIKRLKISSPVRVCQNCYYNLQHERGSEDGPRNCEspecificaciones
AAH15214 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.94kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALFY/KIAA0993/MGC16461/ZFYVE25 | |
WDFY3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23001 | |
WDFY3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGTSKEKAKTATLKQALLGHTDTVTCATASLAYHIIVSGSRDRTCIIWDLNKLSFLTQLRGHRAPVSALCINELTGDIVSCAGTYIHVWSINGNPIVSVNTFTGRSQQIICCCMSEMNEWDTQNVIVTGHSDGVVRFWRMEFLQVPETPAPEPAEVLEMQEDCPEAQIGQEAQDEDSSDSEADEQSISQDPKDTPSQPSSTSHRPRAASCRATAAWCTDSGSDDSRRWSDQLSLDEKDGFIFVNYSEGQTRAHLQGPLSHPHPNPIEVRNYSRLKPGYRWERQLVFRSKLTMHTAFDRKDNAHPAEVTALGISKDHSRILVGDSRGRVFSWSVSDQPGRSAADHWVKDEGGDSCSGCSVRFSLTERRHHCRNCGQLFCQKCSRFQSEIKRLKISSPVRVCQNCYYNLQHERGSEDGPRNC | |
RUO | |
WDFY3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |