Learn More
Abnova™ Human VAC14 Partial ORF (NP_060522.3, 714 a.a. - 782 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055697-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Phosphatidylinositol 3,5-bisphosphate (PI(3,5)P2) is a low-abundance signaling molecule. A regulatory complex made up of VAC14 and FIG4 (MIM 609390) control synthesis of PI(3,5)P2 by activating PI(3)P kinase, FAB1 (PIP5K3; MIM 609414). The VAC14/FIG4 complex also functions in the breakdown of PI(3,5)P2 (Zhang et al., 2007 [PubMed 17956977]).[supplied by OMIM]
Sequence: SHRLQCVPNPELLQTEDSLKAAPKSQKADSPSIDYAELLQHFEKVQNKHLEVRHQRSGRGDHLDRRVVLEspecificaciones
NP_060522.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.33kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SHRLQCVPNPELLQTEDSLKAAPKSQKADSPSIDYAELLQHFEKVQNKHLEVRHQRSGRGDHLDRRVVL | |
RUO | |
VAC14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55697 | |
VAC14 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ArPIKfyve/FLJ10305/FLJ36622/FLJ46582/MGC149815/MGC149816/TAX1BP2/TRX | |
VAC14 | |
Recombinant | |
wheat germ expression system |