missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human UPRT Full-length ORF (NP_659489.1, 1 a.a. - 309 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00139596-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MATELQCPDSMPCHNQQVNSASTPSPEQLRPGDLILDHAGGNRASRAKVILLTGYAHSSLPAELDSGACGGSSLNSEGNSGSGDSSSYDAPAGNSFLEDCELSRQIGAQLKLLPMNDQIRELQTIIRDKTASRGDFMFSADRLIRLVVEEGLNQLPYKECMVTTPTGYKYEGVKFEKGNCGVSIMRSGEAMEQGLRDCCRSIRIGKILIQSDEETQRAKVYYAKFPPDIYRRKVLLMYPILSTGNTVIEAVKVLIEHGVQPSVIILLSLFSTPHGAKSIIQEFPEITILTTEVHPVAPTHFGQKYFGTDEspecificaciones
NP_659489.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781E1243/FUR1/MGC23937/UPP | |
UPRT | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
139596 | |
UPRT (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATELQCPDSMPCHNQQVNSASTPSPEQLRPGDLILDHAGGNRASRAKVILLTGYAHSSLPAELDSGACGGSSLNSEGNSGSGDSSSYDAPAGNSFLEDCELSRQIGAQLKLLPMNDQIRELQTIIRDKTASRGDFMFSADRLIRLVVEEGLNQLPYKECMVTTPTGYKYEGVKFEKGNCGVSIMRSGEAMEQGLRDCCRSIRIGKILIQSDEETQRAKVYYAKFPPDIYRRKVLLMYPILSTGNTVIEAVKVLIEHGVQPSVIILLSLFSTPHGAKSIIQEFPEITILTTEVHPVAPTHFGQKYFGTD | |
RUO | |
UPRT | |
Wheat Germ (in vitro) | |
GST | |
Liquid |