Learn More
ACROBiosystems Human uPAR / PLAUR Protein, His Tag
Marca: ACROBiosystems UPR-H5226-200UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Urokinase plasminogen activator surface receptor (U-PAR) is also known as PLAUR, Monocyte activation antigen Mo3, CD antigen CD87. PLAUR contains three UPAR/Ly6 domains. U-PAR is expressed in neurons of the rolandic area of the brain (at protein level) and is also expressed in the brain. PLAUR / CD87 interacts with MRC2, SRPX2 and SORL1. PLAUR / UPAR acts as a receptor for urokinase plasminogen activator and plays a role in localizing and promoting plasmin formation. U-PAR mediates the proteolysis-independent signal transduction activation effects of U-PA.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 32.3 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AAH02788 | |
200 μg | |
Leu 23 - Arg 303 | |
uPAR,PLAUR,CD87,MO3 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |
32.3 kDa | |
Human uPAR, His Tag (UPR-H5226) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession # AAH02788). | |
-20°C | |
uPAR | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |