missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human UBR2 Full-length ORF (AAH64512.1, 1 a.a. - 439 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00023304-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16167142

  • 528.00€ / 25 microgramos
Fecha estimada de envío: 03-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Proteolysis by the ubiquitin-proteasome system controls the concentration of many regulatory proteins. The selectivity of ubiquitylation is determined by ubiquitin E3 ligases, which recognize the substrate's destabilization signal, or degron. The E3 ligase UBR2 participates in the N-end rule pathway, which targets proteins bearing an N-terminal degron, or N-degron (Kwon et al., 2003 [PubMed 14585983]).[supplied by OMIM]

Sequence: MASELEPEVQAIDRSLLECSAEEIAGKWLQATDLTREVYQHLAHYVPKIYCRGPNPFPQKEDMLAQHVLLGPMEWYLCGEDPAFGFPKLEQANKPSHLCGRVFKVGEPTYSCRDCAVDPTCVLCMECFLGSIHRDHRYRMTTSGGGGFCDCGDTEAWKEGPYCQKHELNTSEIEEEEDPLVHLSEDVIARTYNIFAITFRYAVEILTWEKESELPADLEMVEKSDTYYCMLFNDEVHTYEQVIYTLQKAVNCTQKEAIGFATTVDRDGRRSVRYGDFQYCEQAKSVIVRNTSRQTKPLKVQVMHSSIVAHQNFGLKLLSWLGSIIGYSDGLRRILCQVGLQEGPDGENSSLVDRLMLSDSKLWKGARSVYHQLFMSSLLMDLKYKKLFAVRFAKNYERLQSDYVTDDHDREFSVADLSVQIFTVPSLFSISAGRSGSPL
Especificaciones

Especificaciones

AAH64512.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
76.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C6orf133/DKFZp686C08114/KIAA0349/MGC71112/RP3-392M17.3/bA49A4.1/dJ242G1.1/dJ392M17.3
UBR2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
23304
UBR2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASELEPEVQAIDRSLLECSAEEIAGKWLQATDLTREVYQHLAHYVPKIYCRGPNPFPQKEDMLAQHVLLGPMEWYLCGEDPAFGFPKLEQANKPSHLCGRVFKVGEPTYSCRDCAVDPTCVLCMECFLGSIHRDHRYRMTTSGGGGFCDCGDTEAWKEGPYCQKHELNTSEIEEEEDPLVHLSEDVIARTYNIFAITFRYAVEILTWEKESELPADLEMVEKSDTYYCMLFNDEVHTYEQVIYTLQKAVNCTQKEAIGFATTVDRDGRRSVRYGDFQYCEQAKSVIVRNTSRQTKPLKVQVMHSSIVAHQNFGLKLLSWLGSIIGYSDGLRRILCQVGLQEGPDGENSSLVDRLMLSDSKLWKGARSVYHQLFMSSLLMDLKYKKLFAVRFAKNYERLQSDYVTDDHDREFSVADLSVQIFTVPSLFSISAGRSGSPL
RUO
UBR2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales