missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human UBE2O Partial ORF (NP_071349.2, 361 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00063893-Q01.25ug
Detalles adicionales : Peso : 0.02000kg
Descripción
Sequence: VEPAKIAWECPEKNCAQGEGSMAKKVKRLLKKQVVRIMSCSPDTQCSRDHSMEDPDKKGESKTKSEAESASPEETPDGSASPVEMQDEGAEEPHEAGEQLPPFLLKEGRDEspecificaciones
NP_071349.2 | |
Liquid | |
63893 | |
UBE2O (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
E2-230K/FLJ12878/KIAA1734 | |
UBE2O | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VEPAKIAWECPEKNCAQGEGSMAKKVKRLLKKQVVRIMSCSPDTQCSRDHSMEDPDKKGESKTKSEAESASPEETPDGSASPVEMQDEGAEEPHEAGEQLPPFLLKEGRD | |
RUO | |
UBE2O | |
Wheat Germ (in vitro) | |
GST |