Learn More
Abnova™ Human UBE1DC1 Full-length ORF (NP_079094.1, 1 a.a. - 404 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00079876-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the E1-like ubiquitin-activating enzyme family. This protein activates ubiquitin-fold modifier 1, a ubiquitin-like post-translational modifier protein, via the formation of a high-energy thioester bond. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. A pseudogene located on chromosome 1 has also been identified. [provided by RefSeq]
Sequence: MAESVERLQQRVQELERELAQERSLQVPRSGDGGGGRVRIEKMSSEVVDSNPYSRLMALKRMGIVSDYEKIRTFAVAIVGVGGVGSVTAEMLTRCGIGKLLLFDYDKVELANMNRLFFQPHQAGLSKVQAAEHTLRNINPDVLFEVHNYNITTVENFQHFMDRISNGGLEEGKPVDLVLSCVDNFEARMTINTACNELGQTWMESGVSENAVSGHIQLIIPGESACFACAPPLVVAANIDEKTLKREGVCAASLPTTMGVVAGILVQNVLKFLLNFGTVSFYLGYNAMQDFFPTMSMKPNPQCDDRNCRKQQEEYKKKVAALPKQEVIQEEEEIIHEDNEWGIELVSEVSEEELKNFSGPVPDLPEGITVAYTIPKKQEDSVTELTVEDSGESLEDLMAKMKNMEspecificaciones
NP_079094.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ23251/FLJ23251UBA5/THIFP1/UBE1DC1 | |
UBA5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
79876 | |
UBE1DC1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAESVERLQQRVQELERELAQERSLQVPRSGDGGGGRVRIEKMSSEVVDSNPYSRLMALKRMGIVSDYEKIRTFAVAIVGVGGVGSVTAEMLTRCGIGKLLLFDYDKVELANMNRLFFQPHQAGLSKVQAAEHTLRNINPDVLFEVHNYNITTVENFQHFMDRISNGGLEEGKPVDLVLSCVDNFEARMTINTACNELGQTWMESGVSENAVSGHIQLIIPGESACFACAPPLVVAANIDEKTLKREGVCAASLPTTMGVVAGILVQNVLKFLLNFGTVSFYLGYNAMQDFFPTMSMKPNPQCDDRNCRKQQEEYKKKVAALPKQEVIQEEEEIIHEDNEWGIELVSEVSEEELKNFSGPVPDLPEGITVAYTIPKKQEDSVTELTVEDSGESLEDLMAKMKNM | |
RUO | |
UBA5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |