Learn More
Abnova™ Human TULP2 Partial ORF (NP_003314, 141 a.a. - 250 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007288-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
TULP2 is a member of a family of tubby-like genes (TULPs) that encode proteins of unknown function. Members of this family have been identified in plants, vertebrates, and invertebrates. The TULP proteins share a conserved C-terminal region of approximately 200 amino acid residues. [provided by RefSeq]
Sequence: EVSVENGSVSPPPFKQSPRIRRKGWQAHQRPGTRAEGESDSQDMGDAHKSPNMGPNPGMDGDCVYENLAFQKEEDLEKKREASESTGTNSSAAHNEELSKALKGEGGTDSEspecificaciones
NP_003314 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EVSVENGSVSPPPFKQSPRIRRKGWQAHQRPGTRAEGESDSQDMGDAHKSPNMGPNPGMDGDCVYENLAFQKEEDLEKKREASESTGTNSSAAHNEELSKALKGEGGTDS | |
RUO | |
TULP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7288 | |
TULP2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TUBL2 | |
TULP2 | |
Recombinant | |
wheat germ expression system |