missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TTF2 Partial ORF (NP_003585, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00008458-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16168585

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 13-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the SWI2/SNF2 family of proteins, which play a critical role in altering protein-DNA interactions. The encoded protein has been shown to have dsDNA-dependent ATPase activity and RNA polymerase II termination activity. This protein interacts with cell division cycle 5-like, associates with human splicing complexes, and plays a role in pre-mRNA splicing. [provided by RefSeq]

Sequence: EEVRCPEHGTFCFLKTGVRDGPNKGKSFYVCRADTCSFVRATDIPVSHCLLHEDFVVELQGLLLPQDKKEYRLFFRCIRSKAEGKRWCGSIPWQDPDSK
Especificaciones

Especificaciones

NP_003585
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EEVRCPEHGTFCFLKTGVRDGPNKGKSFYVCRADTCSFVRATDIPVSHCLLHEDFVVELQGLLLPQDKKEYRLFFRCIRSKAEGKRWCGSIPWQDPDSK
RUO
TTF2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8458
TTF2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HuF2
TTF2
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales