missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TSC22D2 Partial ORF (NP_055594, 3 a.a. - 85 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009819-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: KMPAKKKSCFQITSVTTAQVATSITEDTESLDDPDESRTEDVSSEIFDVSRATDYGPEEVCERSSSEETLNNVGDAETPGTVSEspecificaciones
NP_055594 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.87kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KMPAKKKSCFQITSVTTAQVATSITEDTESLDDPDESRTEDVSSEIFDVSRATDYGPEEVCERSSSEETLNNVGDAETPGTVS | |
RUO | |
TSC22D2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9819 | |
TSC22D2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0669/TILZ4a/TILZ4b/TILZ4c | |
TSC22D2 | |
Recombinant | |
wheat germ expression system |