missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRPC6 Partial ORF (NP_004612, 543 a.a. - 592 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00007225-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16166985

  • 349.00€ / 10 microgramos
Fecha estimada de envío: 07-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene forms a receptor-activated calcium channel in the cell membrane. The channel is activated by diacylglycerol and is thought to be under the control of a phosphatidylinositol second messenger system. Activation of this channel occurs independently of protein kinase C and is not triggered by low levels of intracellular calcium. Defects in this gene are a cause of focal segmental glomerulosclerosis 2 (FSGS2). [provided by RefSeq]

Sequence: ARFMAFWHASKAQSIIDANDTLKDLTKVTLGDNVKYYNLARIKWDPSDPQ
Especificaciones

Especificaciones

NP_004612
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.24kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ARFMAFWHASKAQSIIDANDTLKDLTKVTLGDNVKYYNLARIKWDPSDPQ
RUO
TRPC6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7225
TRPC6 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ11098/FLJ14863/FSGS2/TRP6
TRPC6
Recombinant
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales