Learn More
Abnova™ Human TRMU Full-length ORF (AAH80631.1, 1 a.a. - 222 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055687-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is a member of the trmU family. It encodes a mitochondria-specific tRNA-modifying enzyme that is required for the 2-thio modification of 5-taurinomethyl-2-thiouridine tRNA-Lys on the wobble position of the anticodon. [provided by RefSeq]
Sequence: MKKSLSRSTLRSPNGFSEIGLKLEMVSQDALRRTIFPLGGLTKEFVKKIAAENRLHHVLQKKESMGMCFIGKRNFEHFLLQYLQPRPGHFISIEDNKVLGTHKGWFLYTLGQRANIGGLREPWYVVEKDSVKGDVFVAPRTDHPALYRDLLRTSRVHWIAEEPPAALVRDKMMECHFRFRHQMALVCCVLQGGRVPGQREDPAAGAVCLHAPEGPAQSWDGHEspecificaciones
AAH80631.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
51.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC99627/MTO2/MTU1/TRMT/TRMT1/TRNT1 | |
TRMU | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55687 | |
TRMU (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKKSLSRSTLRSPNGFSEIGLKLEMVSQDALRRTIFPLGGLTKEFVKKIAAENRLHHVLQKKESMGMCFIGKRNFEHFLLQYLQPRPGHFISIEDNKVLGTHKGWFLYTLGQRANIGGLREPWYVVEKDSVKGDVFVAPRTDHPALYRDLLRTSRVHWIAEEPPAALVRDKMMECHFRFRHQMALVCCVLQGGRVPGQREDPAAGAVCLHAPEGPAQSWDGH | |
RUO | |
TRMU | |
Wheat Germ (in vitro) | |
GST | |
Liquid |