Learn More
Abnova™ Human TRIM10 Full-length ORF (AAH93926.1, 1 a.a. - 395 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010107-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to cytoplasmic bodies. Studies in mice suggest that this protein plays a role in terminal differentiation of erythroid cells. Alternate splicing of this gene generates two transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MASAASVTSLADEVNCPICQGTLREPVTIDCGHNFCRACLTRYCEIPGPDLEESPTCPLCKEPFRPGSFRPNWQLANVVENIERLQLVSTLGLGEEDVCQEHGEKIYFFCEDDEMQLCVVCREAGEHATHTMRFLEDAAAPYREQIHKCLKCLRKEREEIQEIQSRENKRMQVLLTQVSTKRQQVISEFAHLRKFLEEQQSILLAQLESQDGDILRQRDEFDLLVAGEICRFSALIEELEEKNERPARELLTDIRSTLIRCETRKCRKPVAVSPELGQRIRDFPQQALPLQREMKMFLEKLCFELDYEPAHISLDPQTSHPKLLLSEDHQRAQFSYKWQNSPDNPQRFDRATCVLAHTGITGGRHTWVWMARVPGDSGCCQFCSPPSVLGTEVAAEspecificaciones
AAH93926.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HERF1/MGC141979/RFB30/RNF9 | |
TRIM10 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10107 | |
TRIM10 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASAASVTSLADEVNCPICQGTLREPVTIDCGHNFCRACLTRYCEIPGPDLEESPTCPLCKEPFRPGSFRPNWQLANVVENIERLQLVSTLGLGEEDVCQEHGEKIYFFCEDDEMQLCVVCREAGEHATHTMRFLEDAAAPYREQIHKCLKCLRKEREEIQEIQSRENKRMQVLLTQVSTKRQQVISEFAHLRKFLEEQQSILLAQLESQDGDILRQRDEFDLLVAGEICRFSALIEELEEKNERPARELLTDIRSTLIRCETRKCRKPVAVSPELGQRIRDFPQQALPLQREMKMFLEKLCFELDYEPAHISLDPQTSHPKLLLSEDHQRAQFSYKWQNSPDNPQRFDRATCVLAHTGITGGRHTWVWMARVPGDSGCCQFCSPPSVLGTEVAA | |
RUO | |
TRIM10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |