missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TRAF3IP1 Partial ORF (NP_056465.2, 592 a.a. - 690 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00026146-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Description
Sequence: TLCKSALPLGKIMDYIQEDVDAMQNELQMWHSENRQHAEALQQEQRITDCAVEPLKAELAELEQLIKDQQDKICAVKANILKNEEKIQKMVYSINLTSRSpecifications
NP_056465.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TLCKSALPLGKIMDYIQEDVDAMQNELQMWHSENRQHAEALQQEQRITDCAVEPLKAELAELEQLIKDQQDKICAVKANILKNEEKIQKMVYSINLTSR | |
RUO | |
TRAF3IP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
26146 | |
TRAF3IP1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434F124/MIP-T3/MIPT3 | |
TRAF3IP1 | |
Recombinant | |
wheat germ expression system |