Learn More
Abnova™ Human TPD52L1 Partial ORF (NP_001003396.1, 60 a.a. - 144 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007164-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the tumor protein D52 (TPD52) family. The encoded protein contains a coiled-coil domain and may form homo- or hetero-dimer with TPD52 family members. The protein is reported to be involved in cell proliferation and calcium signaling. It also interacts with the mitogen-activated protein kinase kinase kinase 5 (MAP3K5/ASK1) and positively regulates MAP3K5-induced apoptosis. Multiple alternatively spliced transcript variants have been observed, but the full-length nature of some variants has not yet been determined. [provided by RefSeq]
Sequence: KERHLVEIKQKLGMNLMNELKQNFSKSWHDMQTTTAYKKTHETLSHAGQKATAAFSNVGTAISKKFGDMSYSIRHSISMPAMRRKEspecificaciones
NP_001003396.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.09kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KERHLVEIKQKLGMNLMNELKQNFSKSWHDMQTTTAYKKTHETLSHAGQKATAAFSNVGTAISKKFGDMSYSIRHSISMPAMRRK | |
RUO | |
TPD52L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7164 | |
TPD52L1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D53/MGC8556/TPD52L2/hD53 | |
TPD52L1 | |
Recombinant | |
wheat germ expression system |