Learn More
Abnova™ Human TNIK Full-length ORF (AAH55427.1, 1 a.a. - 538 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00023043-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Germinal center kinases (GCKs), such as TNIK, are characterized by an N-terminal kinase domain and a C-terminal GCK domain that serves a regulatory function (Fu et al., 1999 [PubMed 10521462]).[supplied by OMIM]
Sequence: MKKVTDYSSSSEESESSEEEEEDGESETHDGTVAVSDIPRLIPTGAPGSNEQYNVGMVGTHGLETSHADSFSGSISREGTLMIRETSGEKKRSGHSDSNGFAGHINLPDLVQQSHSPAGTPTEGLGRVSTHSQEMDSGTEYGMGSSTKASFTPFVDPRVYQTSPTDEDEEDEESSATALFTSELLRQEQAKLNEARKISVVNVNPTNIRPHSDTPEIRKYKKRFNSEILCAALWGVNLLVGTENGLMLLDRSGQGKVYNLINRRRFQQMDVLEGLNVLVTISGKKNKLRVYYLSWLRNRILHNDPEVEKKQGWITVGDLEGCIHYKVVKYERIKFLVIALKNAVEIYAWAPKPYHKFMAFKSFADLQHKPLLVDLTVEEGQRLKVIFGSHTGFHVIDVDSGNSYDIYIPSHIQGNITPHAIVILPKTDGMEMLVCYEDEGVYVNTYGRITKDVVLQWGEMPTSVAYIHSNQIMGWGEKAIEIRSVETGHLDGVFMHKRAQRLKFLCERNDKVFFASVRSGGSSQVFFMTLNRNSMMNWEspecificaciones
AAH55427.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
86.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TNIK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
23043 | |
TNIK (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKKVTDYSSSSEESESSEEEEEDGESETHDGTVAVSDIPRLIPTGAPGSNEQYNVGMVGTHGLETSHADSFSGSISREGTLMIRETSGEKKRSGHSDSNGFAGHINLPDLVQQSHSPAGTPTEGLGRVSTHSQEMDSGTEYGMGSSTKASFTPFVDPRVYQTSPTDEDEEDEESSATALFTSELLRQEQAKLNEARKISVVNVNPTNIRPHSDTPEIRKYKKRFNSEILCAALWGVNLLVGTENGLMLLDRSGQGKVYNLINRRRFQQMDVLEGLNVLVTISGKKNKLRVYYLSWLRNRILHNDPEVEKKQGWITVGDLEGCIHYKVVKYERIKFLVIALKNAVEIYAWAPKPYHKFMAFKSFADLQHKPLLVDLTVEEGQRLKVIFGSHTGFHVIDVDSGNSYDIYIPSHIQGNITPHAIVILPKTDGMEMLVCYEDEGVYVNTYGRITKDVVLQWGEMPTSVAYIHSNQIMGWGEKAIEIRSVETGHLDGVFMHKRAQRLKFLCERNDKVFFASVRSGGSSQVFFMTLNRNSMMNW | |
RUO | |
TNIK | |
Recombinant | |
wheat germ expression system |