missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human THEMIS Full-length ORF (ADR83235.1, 1 a.a. - 641 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00387357-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16170454

  • 349.00€ / 10 microgramos
Fecha estimada de envío: 07-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a protein that plays a regulatory role in both positive and negative T-cell selection during late thymocyte development. The protein functions through T-cell antigen receptor signaling, and is necessary for proper lineage commitment and maturation of T-cells. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MALSLEEFVHSLDLRTLPRVLEIQAGIYLEGSIYEMFGNECCFSTGEVIKITGLKVKKIIAEICEQIEGCESLQPFELPMNFPGLFKIVADKTPYLTMEEITRTIHIGPSRLGHPCFYHQKDIKLENLIIKQGEQIMLNSVEEIDGEIMVSCAVARNHQTHSFNLPLSQEGEFYECEDERIYTLKEIVEWKIPKNRTRTVNLTDFSNKWDSTNPFPKDFYGTLILKPVYEIQGVMKFRKDIIRILPSLDVEVKDITDSYDANWFLQLLSTEDLFEMTSKEFPIVTEVIEAPEGNHLPQSILQPGKTIVIHKKYQASRILASEIRSNFPKRHFLIPTSYKGKFKRRPREFPTAYDLEIAKSEKEPLHVVATKAFHSPHDKLSSVSVGDQFLVHQSETTEVLCEGIKKVVNVLACEKILKKSYEAALLPLYMEGGFVEVIHDKKQYPISELCKQFRLPFNVKVSVRDLSIEEDVLAATPGLQLEEDITDSYLLISDFANPTECWEIPVGRLNMTVQLVSNFSRDAEPFLVRTLVEEITEEQYYMMRRYESSASHPPPRPPKHPSVEETKLTLLTLAEERTVDLPKSPKRHHVDITKKLHPNQAGLDSKVLIGSQNDLVDEEKERSNRGATAVAETFKNEKHQK
Especificaciones

Especificaciones

ADR83235.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
97kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C6orf207/FLJ40584/MGC163388/bA325O24.3/bA325O24.4
THEMIS
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
387357
THEMIS (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MALSLEEFVHSLDLRTLPRVLEIQAGIYLEGSIYEMFGNECCFSTGEVIKITGLKVKKIIAEICEQIEGCESLQPFELPMNFPGLFKIVADKTPYLTMEEITRTIHIGPSRLGHPCFYHQKDIKLENLIIKQGEQIMLNSVEEIDGEIMVSCAVARNHQTHSFNLPLSQEGEFYECEDERIYTLKEIVEWKIPKNRTRTVNLTDFSNKWDSTNPFPKDFYGTLILKPVYEIQGVMKFRKDIIRILPSLDVEVKDITDSYDANWFLQLLSTEDLFEMTSKEFPIVTEVIEAPEGNHLPQSILQPGKTIVIHKKYQASRILASEIRSNFPKRHFLIPTSYKGKFKRRPREFPTAYDLEIAKSEKEPLHVVATKAFHSPHDKLSSVSVGDQFLVHQSETTEVLCEGIKKVVNVLACEKILKKSYEAALLPLYMEGGFVEVIHDKKQYPISELCKQFRLPFNVKVSVRDLSIEEDVLAATPGLQLEEDITDSYLLISDFANPTECWEIPVGRLNMTVQLVSNFSRDAEPFLVRTLVEEITEEQYYMMRRYESSASHPPPRPPKHPSVEETKLTLLTLAEERTVDLPKSPKRHHVDITKKLHPNQAGLDSKVLIGSQNDLVDEEKERSNRGATAVAETFKNEKHQK
RUO
THEMIS
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales