Learn More
Abnova™ Human THEM4 Full-length ORF (AAH65277.1, 1 a.a. - 240 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00117145-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Protein kinase B (PKB) is a major downstream target of receptor tyrosine kinases that signal via phosphatidylinositol 3-kinase. Upon cell stimulation, PKB is translocated to the plasma membrane, where it is phosphorylated in the C-terminal regulatory domain. The protein encoded by this gene negatively regulates PKB activity by inhibiting phosphorylation. Transcription of this gene is commonly downregulated in glioblastomas. [provided by RefSeq]
Sequence: MLRSCAARLRTLGALCRPPVGRRLPGSEPRPELRSFSSEEVILKDCSVPNPSWNKDLRLLFDQFMKKCEDGSWKRLPSYKRTPTEWIQDFKTHFLDPKLMKEEQMSQAQLFTRSFDDGLGFEYVMFYNDIEKRMVCLFQGGPYLEGPPGFIHGGAIATMIDATVGMCAMMAGGIVMTANLNINYKRPIPLCSVVMINSQLDKVEGRKFFVSCNVQSVDEKTLYSEATSLFIKLNPAKSLTEspecificaciones
AAH65277.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
53.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CTMP/MGC29636 | |
THEM4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
117145 | |
THEM4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLRSCAARLRTLGALCRPPVGRRLPGSEPRPELRSFSSEEVILKDCSVPNPSWNKDLRLLFDQFMKKCEDGSWKRLPSYKRTPTEWIQDFKTHFLDPKLMKEEQMSQAQLFTRSFDDGLGFEYVMFYNDIEKRMVCLFQGGPYLEGPPGFIHGGAIATMIDATVGMCAMMAGGIVMTANLNINYKRPIPLCSVVMINSQLDKVEGRKFFVSCNVQSVDEKTLYSEATSLFIKLNPAKSLT | |
RUO | |
THEM4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |