Learn More
Abnova™ Human TGM7 Partial ORF (NP_443187, 466 a.a. - 565 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00116179-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Transglutaminases (TGM; EC 2.3.2.13) are a family of structurally and functionally related enzymes that stabilize protein assemblies through the formation of gamma-glutamyl-epsilon lysine crosslinks. For additional background information on transglutaminases, see TGM1 (MIM 190195).[supplied by OMIM]
Sequence: MLGPQRASLPFLDLLESGGLRDQPAQLQLHLARIPEWGQDLQLLLRIQRVPDSTHPRGPIGLVVRFCAQALLHGGGTQKPFWRHTVRMNLDFGKETQWPLEspecificaciones
NP_443187 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLGPQRASLPFLDLLESGGLRDQPAQLQLHLARIPEWGQDLQLLLRIQRVPDSTHPRGPIGLVVRFCAQALLHGGGTQKPFWRHTVRMNLDFGKETQWPL | |
RUO | |
TGM7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
116179 | |
TGM7 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TGMZ | |
TGM7 | |
Recombinant | |
wheat germ expression system |