Learn More
Abnova™ Human TEKT2 Full-length ORF (NP_055281.2, 1 a.a. - 430 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00027285-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. This gene is expressed in the testis and its protein is localized to the flagella of the sperms, indicating that it may play a role in spermatogenesis. [provided by RefSeq]
Sequence: MATLSVKPSRRFQLPDWHTNSYLLSTNAQLQRDASHQIRQEARVLRNETNNQTIWDEHDNRTRLVERIDTVNRWKEMLDKCLTDLDAEIDALTQMKESAEQNLQAKNLPLDVAIECLTLRESRRDIDVVKDPVEDELHKEVEVIEATKKALQQKVSQAFEQLCLLQEVQQQLNSDHRGKMETLEIDRGCLSLNLRSPNISLKVDPTRVPDGSTTLQQWDDFSRFNKDRAEAEMKAATELREATALTIAETNNELEAQRVATEFAFRKRLREMEKVYSELKWQEKNTLEEIAELQEDIRHLEEDLRTKLLSLKLSHTRLEARTYRPNVELCRDQAQYGLTDEVHQLEATIAALKQKLAQAQDALDALCKHLARLQADIACKANSMLLDTKCMDTRRKLTVPAERFVPEVDTFTRTTNSTLSPLKSCQLELAEspecificaciones
NP_055281.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TEKTB1/TEKTIN-T/h-tektin-t | |
TEKT2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
27285 | |
TEKT2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATLSVKPSRRFQLPDWHTNSYLLSTNAQLQRDASHQIRQEARVLRNETNNQTIWDEHDNRTRLVERIDTVNRWKEMLDKCLTDLDAEIDALTQMKESAEQNLQAKNLPLDVAIECLTLRESRRDIDVVKDPVEDELHKEVEVIEATKKALQQKVSQAFEQLCLLQEVQQQLNSDHRGKMETLEIDRGCLSLNLRSPNISLKVDPTRVPDGSTTLQQWDDFSRFNKDRAEAEMKAATELREATALTIAETNNELEAQRVATEFAFRKRLREMEKVYSELKWQEKNTLEEIAELQEDIRHLEEDLRTKLLSLKLSHTRLEARTYRPNVELCRDQAQYGLTDEVHQLEATIAALKQKLAQAQDALDALCKHLARLQADIACKANSMLLDTKCMDTRRKLTVPAERFVPEVDTFTRTTNSTLSPLKSCQLELA | |
RUO | |
TEKT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |