missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TCL1A Partial ORF (NP_068801.1, 61 a.a. - 114 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008115-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: PMTPTQIGPSLLPIMWQLYPDGRYRSSDSSFWRLVYHIKIDGVEDMLLELLPDDEspecificaciones
NP_068801.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.68kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PMTPTQIGPSLLPIMWQLYPDGRYRSSDSSFWRLVYHIKIDGVEDMLLELLPDD | |
RUO | |
TCL1A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8115 | |
TCL1A (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TCL1 | |
TCL1A | |
Recombinant | |
wheat germ expression system |